Learn More
Grainger Flex Metal Conduit Steel Trd Sz 3/4in
Catalog No. 502765165
Encompass_Preferred
The Flexible Metal Conduit Reduced is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made of galvanized steel and has an inner diameter of 1/2 inch. The conduit features interlocked construction for flexibility and protection. It can be used in Class I, Division 2 hazardous locations and is UL listed. The conduit comes in a 10 foot length and has an NPT threaded connector on one end to connect to electrical boxes or fittings. It can be used for exposed runs connecting equipment in industrial, commercial and residential applications. The conduit allows for a bend radius of 6 inches.
Catalog No. 50-276-5165 Supplier Grainger Supplier No. 16R043
The Flexible Metal Conduit Reduced is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made of galvanized steel and has an inner diameter of 1/2 inch. The conduit features interlocked construction for flexibility and protection. It can be used in Class I, Division 2 hazardous locations and is UL listed. The conduit comes in a 10 foot length and has an NPT threaded connector on one end to connect to electrical boxes or fittings. It can be used for exposed runs connecting equipment in industrial, commercial and residential applications. The conduit allows for a bend radius of 6 inches.