Learn More
Grainger Flex Metal Conduit Steel Trd Sz 2in
Catalog No. 502765166
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a sunlight resistant and oil resistant jacket and is UL listed. It comes in 10 foot lengths in trade sizes 3/8 inch to 4 inch. The conduit allows for bends while maintaining its liquid tight properties. It can be used for protection of wires and cables in wet or corrosive environments. The PVC jacket resists oils, acids, and sunlight exposure.
Catalog No. 50-276-5166 Supplier Grainger Supplier No. 16R057
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a sunlight resistant and oil resistant jacket and is UL listed. It comes in 10 foot lengths in trade sizes 3/8 inch to 4 inch. The conduit allows for bends while maintaining its liquid tight properties. It can be used for protection of wires and cables in wet or corrosive environments. The PVC jacket resists oils, acids, and sunlight exposure.