Learn More
Grainger Flex Metal Conduit Steel Trd Sz 2in
Catalog No. 502765179
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is oil resistant and sunlight resistant. It bends easily by hand to route around obstacles.
Catalog No. 50-276-5179 Supplier Grainger Supplier No. 16R059
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is oil resistant and sunlight resistant. It bends easily by hand to route around obstacles.