Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1/2in
Catalog No. 502767681
Encompass_Preferred
The West Indigo 16R009 Flex Metal Conduit Steel TRD is a 1/2 inch flexible metal conduit made of galvanized steel. It has an inner diameter of 0.622 inches and can be used to protect and route electrical wiring. The conduit features a spiral construction that allows flexibility while maintaining the strength of steel. It is UL listed and CSA certified. The 16R009 conduit comes in 10 foot lengths and has a temperature range of -20°F to 150°F. It meets Article 348 of the National Electrical Code standards. This quality conduit provides protection and flexibility for electrical wiring installations.
Catalog No. 50-276-7681 Supplier Grainger Supplier No. 16R009
The West Indigo 16R009 Flex Metal Conduit Steel TRD is a 1/2 inch flexible metal conduit made of galvanized steel. It has an inner diameter of 0.622 inches and can be used to protect and route electrical wiring. The conduit features a spiral construction that allows flexibility while maintaining the strength of steel. It is UL listed and CSA certified. The 16R009 conduit comes in 10 foot lengths and has a temperature range of -20°F to 150°F. It meets Article 348 of the National Electrical Code standards. This quality conduit provides protection and flexibility for electrical wiring installations.