Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1 1/2in
Catalog No. 502767683
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a sunlight resistant and oil resistant jacket and is UL listed. It comes in 10 foot lengths in trade sizes 3/8 inch to 4 inch. The conduit allows for bends while maintaining its liquid tight properties. It can be used for protection of wires and cables in wet or damp locations.
Catalog No. 50-276-7683 Supplier Grainger Supplier No. 16R018
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a sunlight resistant and oil resistant jacket and is UL listed. It comes in 10 foot lengths in trade sizes 3/8 inch to 4 inch. The conduit allows for bends while maintaining its liquid tight properties. It can be used for protection of wires and cables in wet or damp locations.