Learn More
Grainger Flex Metal Conduit Steel Trd Sz 3/4in
Catalog No. 502765964
Encompass_Preferred
The Flexible Metal Conduit Reduced is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing flexibility. This 3/4" trade size conduit measures 7/8" OD and is available in 25' lengths. It can be used with standard 3/4" fittings and is rated for both indoor and outdoor applications. This UL listed conduit offers good physical protection for cables and wires in exposed runs.
Catalog No. 50-276-5964 Supplier Grainger Supplier No. 16R051
The Flexible Metal Conduit Reduced is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing flexibility. This 3/4" trade size conduit measures 7/8" OD and is available in 25' lengths. It can be used with standard 3/4" fittings and is rated for both indoor and outdoor applications. This UL listed conduit offers good physical protection for cables and wires in exposed runs.