Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1/2in
Catalog No. 502768505
Encompass_Preferred
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing it to be bent and shaped easily. This 3/4" trade size conduit can be used with 3/8" to 1/2" actual size conductors. It is rated for both indoor and outdoor applications. This UL listed product meets Article 348 of the National Electrical Code. The conduit comes in a 10 foot length and has a temperature range of -20°F to 150°F. It provides good physical protection for cables and wires.
Catalog No. 50-276-8505 Supplier Grainger Supplier No. 16R031
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing it to be bent and shaped easily. This 3/4" trade size conduit can be used with 3/8" to 1/2" actual size conductors. It is rated for both indoor and outdoor applications. This UL listed product meets Article 348 of the National Electrical Code. The conduit comes in a 10 foot length and has a temperature range of -20°F to 150°F. It provides good physical protection for cables and wires.