Learn More
Grainger Flex Metal Conduit Steel Trd Sz 2in
Catalog No. 502768501
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is corrosion, moisture, and crush resistant. It meets NEC code requirements for an exposed run conduit.
Catalog No. 50-276-8501 Supplier Grainger Supplier No. 16R019
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is corrosion, moisture, and crush resistant. It meets NEC code requirements for an exposed run conduit.