Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1 1/2in
Catalog No. 502767686
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a UL and CSA listing and is sunlight and oil resistant. It comes in lengths of 25 ft, 50 ft, and 100 ft with trade sizes of 1/2 in, 3/4 in, 1 in, 1-1/4 in, 1-1/2 in, and 2 in. The conduit allows for a smaller bend radius than standard metal conduit. It can be used for connections to motors, transformers, and other electrical equipment where flexibility and sealing against liquids and vapors is required. The PVC jacket resists oil, sunlight, moisture and abrasion.
Catalog No. 50-276-7686 Supplier Grainger Supplier No. 16R056
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a UL and CSA listing and is sunlight and oil resistant. It comes in lengths of 25 ft, 50 ft, and 100 ft with trade sizes of 1/2 in, 3/4 in, 1 in, 1-1/4 in, 1-1/2 in, and 2 in. The conduit allows for a smaller bend radius than standard metal conduit. It can be used for connections to motors, transformers, and other electrical equipment where flexibility and sealing against liquids and vapors is required. The PVC jacket resists oil, sunlight, moisture and abrasion.