Learn More
Grainger Flex Metal Conduit Steel Trd Sz 3/4in
Catalog No. 502768357
Encompass_Preferred
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for wires and cables in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing flexibility. This 3/4 in. trade size conduit can be used with a variety of fittings and connectors. It is rated for both indoor and outdoor use. The working temperature range is -20°F to 140°F. This UL listed conduit meets Article 348 of the National Electrical Code standards. The conduit is sold by the foot and is available in lengths up to 100 ft.
Catalog No. 50-276-8357 Supplier Grainger Supplier No. 16R013
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for wires and cables in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing flexibility. This 3/4 in. trade size conduit can be used with a variety of fittings and connectors. It is rated for both indoor and outdoor use. The working temperature range is -20°F to 140°F. This UL listed conduit meets Article 348 of the National Electrical Code standards. The conduit is sold by the foot and is available in lengths up to 100 ft.