Learn More
Grainger Flex Metal Conduit Steel Trd Sz 3/4in
Catalog No. 502768358
Encompass_Preferred
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing flexibility. This 3/4" trade size conduit can be used with 3/8" to 1/2" conductors. It is rated for both indoor and outdoor use. This UL listed conduit bends easily by hand to route around obstacles. It comes in 10' lengths and has an oil resistant gray enamel finish.
Catalog No. 50-276-8358 Supplier Grainger Supplier No. 16R034
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing flexibility. This 3/4" trade size conduit can be used with 3/8" to 1/2" conductors. It is rated for both indoor and outdoor use. This UL listed conduit bends easily by hand to route around obstacles. It comes in 10' lengths and has an oil resistant gray enamel finish.