Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1/2in
Catalog No. 502768447
Encompass_Preferred
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing it to be bent and shaped easily. This 3/4" trade size conduit can be used with 3/8" to 1/2" conductors and reduces down to 1/2" trade size. It is UL listed and CSA certified. The working temperature range is between -20°F to 140°F. This conduit comes in a 10 ft length and has a gray finish. It can be used for exposed runs, connections to motors, and protection in damp or wet locations.
Catalog No. 50-276-8447 Supplier Grainger Supplier No. 16R008
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It is made from galvanized steel and has an inner corrugated surface allowing it to be bent and shaped easily. This 3/4" trade size conduit can be used with 3/8" to 1/2" conductors and reduces down to 1/2" trade size. It is UL listed and CSA certified. The working temperature range is between -20°F to 140°F. This conduit comes in a 10 ft length and has a gray finish. It can be used for exposed runs, connections to motors, and protection in damp or wet locations.