Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1in
Catalog No. 502768451
Encompass_Preferred
The Grainger 16R016 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It can be used with pumps, fans, blowers, conveyors, compressors, and other industrial equipment. The motor dimensions are 9-1/2" long x 7-3/4" wide x 8-7/8" high and it weighs approximately 36 pounds.
Catalog No. 50-276-8451 Supplier Grainger Supplier No. 16R016
The Grainger 16R016 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It can be used with pumps, fans, blowers, conveyors, compressors, and other industrial equipment. The motor dimensions are 9-1/2" long x 7-3/4" wide x 8-7/8" high and it weighs approximately 36 pounds.