Learn More
Opentrons Labworks Inc FLEX NGS WORKSTATION 2X8CHANNL
Catalog No. NC3779164
Encompass
  • Automates Next-Generation Sequencing workflows
  • Enhances efficiency and reduces errors
  • Minimizes hands-on time
  • Configured with two 8-channel pipettes for library preparation
  • Supports automation for up to 48 samples
  • Includes Flex 8-channel pipettes of 1-50 µL and 5-1000 µL
  • Equipped with Flex Gripper GEN1
  • Includes Flex Magnetic Block, Temperature Module, and Thermocycler
  • Offers Wi-Fi, Ethernet, and USB connectivity
  • Open-source Python API and code-free Protocol Designer software
  • Controlled via touchscreen and Opentrons App
  • Dimensions: 870 mm width, 690 mm depth, 840 mm height
  • Requires 20 cm clearance for installation
  • 1-year factory warranty
  • On-site installation required
Catalog No. NC3779164 Supplier Opentrons Labworks Inc Supplier No. 99100354
Add to Cart
Add to Cart
Request a Quote
  • Automates Next-Generation Sequencing workflows
  • Enhances efficiency and reduces errors
  • Minimizes hands-on time
  • Configured with two 8-channel pipettes for library preparation
  • Supports automation for up to 48 samples
  • Includes Flex 8-channel pipettes of 1-50 µL and 5-1000 µL
  • Equipped with Flex Gripper GEN1
  • Includes Flex Magnetic Block, Temperature Module, and Thermocycler
  • Offers Wi-Fi, Ethernet, and USB connectivity
  • Open-source Python API and code-free Protocol Designer software
  • Controlled via touchscreen and Opentrons App
  • Dimensions: 870 mm width, 690 mm depth, 840 mm height
  • Requires 20 cm clearance for installation
  • 1-year factory warranty
  • On-site installation required