Learn More
Grainger Starrett Indicator Holding Rod, 3 in. Length, 3/8 in. Rod Diameter, 1 ea.
Catalog No. 502786557
Encompass_Preferred
The Flex-O-Post Indicator Holder is a precision component designed for accurate positioning and alignment of measuring instruments in machining and calibration applications. This individual component serves as an indicator holding rod for specified holder systems, ensuring stable and reliable performance during setup.
  • Features an adjustable design for precise positioning
  • Constructed from durable materials for long-lasting use
  • Includes fine adjustment controls for accurate measurements
  • Points screw into the handle and fit either end
  • Tapered points enhance visibility of the scriber point on the work
  • Designed with a 0.375 in. (3/8 in.) rod diameter and a 3 in. overall length
Catalog No. 50-278-6557 Supplier Grainger Supplier No. 197T21
The Flex-O-Post Indicator Holder is a precision component designed for accurate positioning and alignment of measuring instruments in machining and calibration applications. This individual component serves as an indicator holding rod for specified holder systems, ensuring stable and reliable performance during setup.
  • Features an adjustable design for precise positioning
  • Constructed from durable materials for long-lasting use
  • Includes fine adjustment controls for accurate measurements
  • Points screw into the handle and fit either end
  • Tapered points enhance visibility of the scriber point on the work
  • Designed with a 0.375 in. (3/8 in.) rod diameter and a 3 in. overall length