Learn More
Biolegend Flex-T™ HLA-A*03:01 Monomer UVX
Supplier:  Biolegend 280005
Encompass_Preferred
Flex-T™ HLA-A*03:01 Monomer UVX Reactivity: Human; Apps: ELISA, FC; Size: 50 ug
Catalog No. 50-207-2585