Learn More
MSC Rubbermaid FLex Dispenser, 500 mL, Plastic, Black, ADA Compliant
Catalog No. 502836806
Encompass
The FLex Dispenser is a manual wall-mounted system designed for efficiency, supporting both liquid and foam soaps. It offers adjustable dose sizes for flexibility and features an antimicrobial push bar for a sanitary washroom experience.
  • Highest yield of hand washes-up to 3,250 per refill.
  • Accurate, adjustable, on-unit dose meter for effective portion control (0.4, 0.65, 1.0 mL target dosage).
  • Fully ADA compliant in operation, force, and depth.
  • Antimicrobial push bar features a specially-treated coating to help reduce the growth of bacteria and germs.
  • Compatible with moisturizing foam hand soap.
Catalog No. 50-283-6806 Supplier MSC Supplier No. 57854770
The FLex Dispenser is a manual wall-mounted system designed for efficiency, supporting both liquid and foam soaps. It offers adjustable dose sizes for flexibility and features an antimicrobial push bar for a sanitary washroom experience.
  • Highest yield of hand washes-up to 3,250 per refill.
  • Accurate, adjustable, on-unit dose meter for effective portion control (0.4, 0.65, 1.0 mL target dosage).
  • Fully ADA compliant in operation, force, and depth.
  • Antimicrobial push bar features a specially-treated coating to help reduce the growth of bacteria and germs.
  • Compatible with moisturizing foam hand soap.