Learn More
MSC Rubbermaid Soap Dispenser, Wall Mounted, 500 mL, White
Catalog No. 502838706
Encompass
This manual wall-mounted dispenser features a revolutionary design that increases facility efficiency by allowing the use of two different soaps (liquid or foam) with a single unit. It offers adjustable dose sizes and is designed with an antimicrobial push bar to prevent bacterial growth, helping to maintain a sanitary washroom experience.
  • Highest yield of hand washes, up to 3,250 per refill.
  • Accurate, adjustable, on-unit dose meter for effective portion control (0.4, 0.65, 1.0 mL target dosage).
  • Superior "clean look" styling improves washroom aesthetics.
  • Fully ADA compliant in terms of operation, force, and depth.
  • Antimicrobial push bar features a specially-treated coating to help reduce the growth of bacteria and germs.
  • Increases productivity by making replacements easier by working interchangeably with either lotion or foam refills.
Catalog No. 50-283-8706 Supplier MSC Supplier No. 57844649
This manual wall-mounted dispenser features a revolutionary design that increases facility efficiency by allowing the use of two different soaps (liquid or foam) with a single unit. It offers adjustable dose sizes and is designed with an antimicrobial push bar to prevent bacterial growth, helping to maintain a sanitary washroom experience.
  • Highest yield of hand washes, up to 3,250 per refill.
  • Accurate, adjustable, on-unit dose meter for effective portion control (0.4, 0.65, 1.0 mL target dosage).
  • Superior "clean look" styling improves washroom aesthetics.
  • Fully ADA compliant in terms of operation, force, and depth.
  • Antimicrobial push bar features a specially-treated coating to help reduce the growth of bacteria and germs.
  • Increases productivity by making replacements easier by working interchangeably with either lotion or foam refills.