Learn More
Nova Biomedical Corporation FLEX2 Autosampler STM Tubing Harness, Automated Sample Aspiration, Durable & Flexible Design, Each
Catalog No. NC3367654
Encompass
The FLEX2 Autosampler STM Tubing Harness is an essential component for the BioProfile FLEX2 On-Line Autosampler system, enhancing automated sample aspiration and transfer in cell culture analysis. Its flexible, durable design ensures compatibility and performance, optimizing system integrity and efficiency for accurate results in laboratory environments. This replacement part supports walkaway automation, allowing uninterrupted operation and maintaining a sterile fluid path.
Catalog No. NC3367654 Supplier Nova Biomedical Corporation Supplier No. 61372
The FLEX2 Autosampler STM Tubing Harness is an essential component for the BioProfile FLEX2 On-Line Autosampler system, enhancing automated sample aspiration and transfer in cell culture analysis. Its flexible, durable design ensures compatibility and performance, optimizing system integrity and efficiency for accurate results in laboratory environments. This replacement part supports walkaway automation, allowing uninterrupted operation and maintaining a sterile fluid path.