Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems Flexacam i5

Catalog No. 01001359 Shop All Leica Microsystems Products
Change view
Click to view available options
For Use With (Equipment):
Compound Microscopes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-001-359 Compound Microscopes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-359 Supplier Leica Microsystems Supplier No. 12730537
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color camera for compound microscopes with high definition live image

  • Digital color camera with CMOS sensor (1/2.3) in.
  • Possible to operate with 4k HDMI monitor using Enersight OSD
  • High definition live image 4k, 60 fps, HDR
  • JPG images with up to 12 MP
  • Full HD MJPG Video recording with 30 fps
  • Camera control: via PC (Windows) using Enersight Desktop, via smartphone / tablet by Enersight Mobile App (connection to Ethernet or Wifi is required) or via USB mouse (not included) (Enersight OSD mode)
  • Capture images directly to USB Stick or send images via E-Mail (Enersight OSD mode)
  • USB-C connection to PC for Enersight Desktop
  • Supported OS: Win 10 Pro/ Win 11 / iOS / Android
  • Delivered accessories: HDMI Cable, USB C-A cable, USB stick
TRUSTED_SUSTAINABILITY

Specifications

Type Camera
CAS 67-56-1, 322-25-8
Concentration 1.0 mg/mL in methanol (as free base)
Packaging Ampule
Linear Formula C10H14FNO · HCl
Quantity 1 mL
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Flash Point 9°C (48.2°F)
For Use With (Equipment) Compound Microscopes
Formula Weight 219.68
Includes HDMI Cable, USB C-A cable, USB stick
Port Type USB C-A
Sensor Types CMOS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.