Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Flexal™ Nitrile Gloves

Catalog No. 19066130 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-066-130 2X-Large
19-066-125 X-Small
19-066-126 Small
19-066-127 Medium
19-066-128 Large
19-066-129 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-066-130 Supplier Cardinal Health™ Supplier No. 88TN06XXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitrile medical gloves provide tactile sensitivity and protect hands from potentially infectious materials.

  • Textured fingertips improve grip even when wet
  • Tested for use with various lab chemicals and chemotherapy drugs
  • Latex-free
  • Powder free
  • Ambidextrous

Specifications

Certifications/Compliance ASTM D6319, ASTM D6978
Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory
AQL 1
Color Blue
Disposable Single-use
Finger Thickness (English) 3.7 mil
Finger Thickness (Metric) 0.09 mm
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 2X-Large
Thickness (English) 3.7 mil
Thickness (Metric) 0.10 mm
FDA 510 (k) Yes
Model 88TN06XXL
Series Flexal™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.