Learn More
MSC Flexbar C-Clamp Flexiguard, 7 1/2 x 10 in, 1 ea
Catalog No. 5000782036
Encompass
The C-Clamp Flexiguard is a safety shield featuring curved plastic shields mounted on rugged, flexible goosenecks. It is designed to positively deflect chips and coolant, allowing for clearer and closer visual inspection of the work being performed. The C-clamp enables the safety shield to be affixed to various locations, including flat or round surfaces.
  • Positively deflects chips and coolant
  • Permits clear and closer visual inspection of work
  • Allows clamping of the safety shield on hard-to-affix locations (flat, round, etc.)
  • Made of malleable iron with 120° vee on the clamp
Catalog No. 50-007-82036 Supplier MSC Supplier No. 02598969
The C-Clamp Flexiguard is a safety shield featuring curved plastic shields mounted on rugged, flexible goosenecks. It is designed to positively deflect chips and coolant, allowing for clearer and closer visual inspection of the work being performed. The C-clamp enables the safety shield to be affixed to various locations, including flat or round surfaces.
  • Positively deflects chips and coolant
  • Permits clear and closer visual inspection of work
  • Allows clamping of the safety shield on hard-to-affix locations (flat, round, etc.)
  • Made of malleable iron with 120° vee on the clamp