Learn More
MSC Flexbar Drill Press Replacement Shield, Wing-Shaped, Lexan, 6 in
Catalog No. 5000782352
Encompass
This is a Wing-Shaped Drill Press Replacement Shield designed to provide protection when using a Drill/Chuck Visorguard. It is constructed from durable Lexan material.
  • Replacement shield
  • Wing-shaped style
  • Lexan material
  • Width: 6 in
  • Length: 6 in
  • For use with drill/chuck visorguard
Catalog No. 50-007-82352 Supplier MSC Supplier No. 01050509
This is a Wing-Shaped Drill Press Replacement Shield designed to provide protection when using a Drill/Chuck Visorguard. It is constructed from durable Lexan material.
  • Replacement shield
  • Wing-shaped style
  • Lexan material
  • Width: 6 in
  • Length: 6 in
  • For use with drill/chuck visorguard