Learn More
MSC Flexbar Indicator Positioner & Holder, Flexible Arm, 150 lb Magnetic Pull, 21 in Overall Length
Catalog No. 5000514134
Encompass
This indicator positioner and holder comes with a flexible arm and a magnetic base. It features a strong 150 lb magnetic pull for secure attachment and has specific base dimensions including a length of 2-3/4 in, a height of 3-1/8 in, and a width of 2-3/4 in, designed to position and hold indicators securely.
  • Flexible arm allows for versatile and precise positioning of indicators.
  • 150 lb magnetic pull ensures strong and secure attachment to ferrous surfaces.
  • Includes base, provided as a complete unit, ready for immediate use.
  • Rectangular magnetic base offers a stable and reliable foundation for various applications.
  • No fine adjustment, suggesting a durable and straightforward design suitable for general positioning tasks.
Catalog No. 50-005-14134 Supplier MSC Supplier No. 60757267
This indicator positioner and holder comes with a flexible arm and a magnetic base. It features a strong 150 lb magnetic pull for secure attachment and has specific base dimensions including a length of 2-3/4 in, a height of 3-1/8 in, and a width of 2-3/4 in, designed to position and hold indicators securely.
  • Flexible arm allows for versatile and precise positioning of indicators.
  • 150 lb magnetic pull ensures strong and secure attachment to ferrous surfaces.
  • Includes base, provided as a complete unit, ready for immediate use.
  • Rectangular magnetic base offers a stable and reliable foundation for various applications.
  • No fine adjustment, suggesting a durable and straightforward design suitable for general positioning tasks.