Learn More
MSC Flexbar Replacement Shield, 7-1/2 in x 10 in, Polycarbonate, 3/Pk.
Catalog No. 5000796219
Encompass
These replacement shields are designed for use with Flexiguard products in machine safety applications. They provide protection against chips and debris, contributing to a safer industrial work environment.
  • Measures 7-1/2 in x 10 in
  • Constructed from polycarbonate
  • Supplied in a pack of 3 shields
  • Suitable for general purpose machine safety applications
Catalog No. 50-007-96219 Supplier MSC Supplier No. 02599025
These replacement shields are designed for use with Flexiguard products in machine safety applications. They provide protection against chips and debris, contributing to a safer industrial work environment.
  • Measures 7-1/2 in x 10 in
  • Constructed from polycarbonate
  • Supplied in a pack of 3 shields
  • Suitable for general purpose machine safety applications