Learn More
MSC Flexbar Replacement Shield, Acrylic, 12 in x 16 in, 3/Pk
Catalog No. 5000770180
Encompass
This replacement safety shield is designed for use with Flexbar Flexiguard systems, offering clear visibility and protection.
  • Made from acrylic material
  • Measures 12 in wide by 16 in long
  • Comes in a package of 3 shields
  • Shield thickness is 1/8 in
  • Interchangeable with other Flexiguard models
Catalog No. 50-007-70180 Supplier MSC Supplier No. 02598977
This replacement safety shield is designed for use with Flexbar Flexiguard systems, offering clear visibility and protection.
  • Made from acrylic material
  • Measures 12 in wide by 16 in long
  • Comes in a package of 3 shields
  • Shield thickness is 1/8 in
  • Interchangeable with other Flexiguard models