Learn More
MSC Flexbar Replacement Shield, Lexan, 16x8x16 in, for Latheguard, 1 ea
Catalog No. 5000789091
Encompass
This replacement shield is constructed from Lexan plastic, providing durable protection for use with Latheguard Shields. It is designed to offer a clear barrier while maintaining visibility during operation.
  • Constructed from Lexan plastic
  • Designed for use with Latheguard Shields
  • Measures 16 in long, 8 in wide, and 16 in high/thick
Catalog No. 50-007-89091 Supplier MSC Supplier No. 06529762
This replacement shield is constructed from Lexan plastic, providing durable protection for use with Latheguard Shields. It is designed to offer a clear barrier while maintaining visibility during operation.
  • Constructed from Lexan plastic
  • Designed for use with Latheguard Shields
  • Measures 16 in long, 8 in wide, and 16 in high/thick