Learn More
Analytical Sales & Services FlexChrom UHPLC SS Flexible Tubing (without fittings) – 30cm x 0.005" ID
Catalog No. 50456301
Encompass
FlexChrom, 1/16 in. Flexible UHPLC Flexible SS Tub (no fittings) 30cm x 0.005 in. (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-456-301 Supplier Analytical Sales & Services Supplier No. 990028FLEX
FlexChrom, 1/16 in. Flexible UHPLC Flexible SS Tub (no fittings) 30cm x 0.005 in. (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)