Learn More
Analytical Sales & Services FlexChrom UHPLC SS Flexible Tubing (without fittings) – 10cm x 0.010" ID
Catalog No. 50456284
Encompass
FlexChrom, 1/16 in. Flexible UHPLC Flexible SS Tub (no fittings) 10cm x 0.010 in. (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-456-284 Supplier Analytical Sales & Services Supplier No. 970011FLEX
FlexChrom, 1/16 in. Flexible UHPLC Flexible SS Tub (no fittings) 10cm x 0.010 in. (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)