Learn More
Analytical Sales & Services FlexChrom Double UHPLC Assembly – 30cm x 0.010" ID
Catalog No. 50456294
Encompass
FlexChrom, Double 1/16 in. Flexible UHPLC Assembly, 30cm x 0.010 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-456-294 Supplier Analytical Sales & Services Supplier No. 970228FLEX
FlexChrom, Double 1/16 in. Flexible UHPLC Assembly, 30cm x 0.010 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)