Learn More
Analytical Sales & Services FlexChrom Double UHPLC Assembly – 30cm x 0.005" ID
Catalog No. 50456318
Encompass
FlexChrom, Double 1/16 in. Flexible UHPLC Assembly, 30cm x 0.005 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-456-318 Supplier Analytical Sales & Services Supplier No. 990228FLEX
FlexChrom, Double 1/16 in. Flexible UHPLC Assembly, 30cm x 0.005 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)