Learn More
Analytical Sales & Services FlexChrom Double UHPLC Assembly - 10cm
Catalog No. 5000805930
Encompass
    \n
  • Pre-finished Stainless Steel
  • \n
  • One-piece design – no brazed-on sleeves
  • \n
  • 1/16″ OD at the ends, 1/32″ OD in the middle
  • \n
  • True ZDV
  • \n
  • Ultra high pressure – successfully tested to 27,000 psi
  • \n
  • Double UHPLC assembly, includes two Stainless Steel fingertight fittings
  • \n
Catalog No. 50-008-05930 Supplier Analytical Sales & Services Supplier No. 990241FLEX3
    \n
  • Pre-finished Stainless Steel
  • \n
  • One-piece design – no brazed-on sleeves
  • \n
  • 1/16″ OD at the ends, 1/32″ OD in the middle
  • \n
  • True ZDV
  • \n
  • Ultra high pressure – successfully tested to 27,000 psi
  • \n
  • Double UHPLC assembly, includes two Stainless Steel fingertight fittings
  • \n