Learn More
Analytical Sales & Services FlexChrom Single Flexible UHPLC Assembly – 20cm x 0.007" ID
Catalog No. 50455916
Encompass
FlexChrom, Single 1/16 in. Flexible UHPLC Assembly, 20cm x 0.007 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-455-916 Supplier Analytical Sales & Services Supplier No. 930115FLEX
FlexChrom, Single 1/16 in. Flexible UHPLC Assembly, 20cm x 0.007 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)