Learn More
Analytical Sales & Services FlexChrom Single UHPLC Assembly – 10cm x 0.005" ID
Catalog No. 50456312
Encompass
FlexChrom, Single 1/16 in. Flexible UHPLC Assembly, 10cm x 0.005 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-456-312 Supplier Analytical Sales & Services Supplier No. 990111FLEX
FlexChrom, Single 1/16 in. Flexible UHPLC Assembly, 10cm x 0.005 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)