Learn More
Analytical Sales & Services FlexChrom Single UHPLC Assembly - 30cm
Catalog No. 5000806911
Encompass
    \n
  • Pre-finished Stainless Steel
  • \n
  • One-piece design – no brazed-on sleeves
  • \n
  • 1/16″ OD at the ends, 1/32″ OD in the middle
  • \n
  • True ZDV
  • \n
  • Ultra high pressure – successfully tested to 27,000 psi
  • \n
  • Single UHPLC assembly, includes one Stainless Steel fingertight fitting
  • \n
Catalog No. 50-008-06911 Supplier Analytical Sales & Services Supplier No. 990128FLEX3
    \n
  • Pre-finished Stainless Steel
  • \n
  • One-piece design – no brazed-on sleeves
  • \n
  • 1/16″ OD at the ends, 1/32″ OD in the middle
  • \n
  • True ZDV
  • \n
  • Ultra high pressure – successfully tested to 27,000 psi
  • \n
  • Single UHPLC assembly, includes one Stainless Steel fingertight fitting
  • \n