Learn More
Analytical Sales & Services FlexChrom UHPLC SS Flexible Tubing (without fittings) - 10cm
Catalog No. 5000804258
Encompass
    \n
  • Pre-finished Stainless Steel
  • \n
  • One-piece design – no brazed-on sleeves
  • \n
  • 1/16″ OD at the ends, 1/32″ OD in the middle
  • \n
  • True ZDV
  • \n
  • Ultra high pressure – successfully tested to 27,000 psi
  • \n
  • Does not include fittings
  • \n
Catalog No. 50-008-04258 Supplier Analytical Sales & Services Supplier No. 930011FLEX3
    \n
  • Pre-finished Stainless Steel
  • \n
  • One-piece design – no brazed-on sleeves
  • \n
  • 1/16″ OD at the ends, 1/32″ OD in the middle
  • \n
  • True ZDV
  • \n
  • Ultra high pressure – successfully tested to 27,000 psi
  • \n
  • Does not include fittings
  • \n