Learn More
AVANTOR MASTERFLEX (VWR LLC) Tubing, Flexelene, 1/2 X 3/4, 50' /Pk
Catalog No. NC0631270
Encompass
Flexelene 135 C Tubing, 1/2 X 3/4, 50' /PkADI- and DEHP-free for the highest level of purity in your process!Flexelene 135C is an ideal tubing for pharmaceutical, biotech, life science, research, and general-purpose lab applications. Translucent, with no plasticizers, latex, or vinyl acetate, Flexelene 135 C is also animal-derived ingredient-free. This flexible tubing is made of non-allergenic materials and offers ultra-low extractables and leachables.
Catalog No. NC0631270 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9618110
Flexelene 135 C Tubing, 1/2 X 3/4, 50' /PkADI- and DEHP-free for the highest level of purity in your process!Flexelene 135C is an ideal tubing for pharmaceutical, biotech, life science, research, and general-purpose lab applications. Translucent, with no plasticizers, latex, or vinyl acetate, Flexelene 135 C is also animal-derived ingredient-free. This flexible tubing is made of non-allergenic materials and offers ultra-low extractables and leachables.