Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Typenex™ Flexi 2.1™ Blood Band
SDP

Catalog No. 22355681
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-355-681 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-355-681 Supplier Typenex™ Supplier No. FLX975
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made of a soft, flexible material. Typenex Flexi 2.1 Blood Band offers the necessary durability along with the comfort required for a long-term stay or outpatient scenario.

  • A barcoded blood bank identification number (BBID) improves patient safety by eliminating data entry errors
  • Easy-to-read prompts for patient information written in a large font that can quickly be read by caregivers
  • Water resistant plastic cover keeps patient information legible throughout the patient’s stay, even with daily showering
  • Two options for forms that best fits your identification protocols

Specifications

Color Blue
Product Type Blood Band
For Use With (Application) Allows blood bank staff to use a scanner to transmit it directly into the Laboratory Information System (LIS).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.