Learn More
Research Products International Corp Flexi-Rack Work Station
SDP
Catalog No. 501977983
Encompass
FlexiRack features a modular design that provides a custom rack based on individual requirements. It accommodates tubes from 0.5-50 ml by selecting the proper color coded insert. Easy release tab allow inserts to be quickly changed depending on the application. Tiered rack style permits full viewing of all tubes and saves valuable bench space.
The complete system includes:
  • two racks
  • one 12x0.5ml insert
  • one 12x1.5ml insert
  • one 4x15ml insert
  • one 3x50ml insert
  • two connection brackets
All components are made of heavy duty polypropylene for rugged durability and can be autoclaved. Extra inserts can be ordered separately.
Catalog No. 50-197-7983 Supplier Research Products International Corp Supplier No. 247515
FlexiRack features a modular design that provides a custom rack based on individual requirements. It accommodates tubes from 0.5-50 ml by selecting the proper color coded insert. Easy release tab allow inserts to be quickly changed depending on the application. Tiered rack style permits full viewing of all tubes and saves valuable bench space.
The complete system includes:
  • two racks
  • one 12x0.5ml insert
  • one 12x1.5ml insert
  • one 4x15ml insert
  • one 3x50ml insert
  • two connection brackets
All components are made of heavy duty polypropylene for rugged durability and can be autoclaved. Extra inserts can be ordered separately.