Learn More
Description
Flexible Collodion USP is a clear, or slightly opalescent, viscous liquid. It is colorless or slightly yellow, and has the odor of ether. The strong odor of camphor becomes noticeable as the ether evaporates.
Specifications
Specifications
| Chemical Name or Material | Flexible Collodion USP |
| Packaging | Bottle, Amber Glass, Boston Round, 500mL, 38-439 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | Unknown |
| Quantity | 500 mL |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
