Learn More
Sigma Aldrich Flexible conductive silver paste for screen printing
Catalog No. 90176925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-176-925G 25 g
90-176-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-176-925G Supplier Sigma Aldrich Supplier No. 90176925G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
This product is a thermoplastic screen printing ink, developed for use in membrane touch switch applications as well as counter electrode in electrochemical sensors and conductive tracks. It may be screen printed on to flexible polymeric substrates, such as polyimide, PEN and PET substrates, to create a highly conductive track for general circuitry. The ink has undergone electrical and mechanical testing which has shown that it is very flexible.Product of Gwent Electronic Materials Ltd (GEM), part of Sun Chemical Corporation
Specifications
Quantity 25 g
Synonym Ag ink; C2080415P2; Conductive ink; Silver ink