Learn More
Grainger Hi-Tech Duravent Industrial Ducting Hose, PVC, 12 in. ID, 25 ft., Clear
Catalog No. NC0209737
Encompass_Preferred
This industrial ducting hose is a flexible, transparent solution designed for the efficient movement of large volumes of air. It features a clear, durable polyvinylchloride (PVC) wall reinforced with a spring wire helix, making it a highly compressible and versatile choice for demanding industrial environments. The hose is well-suited for applications involving chemical fume removal, solvent ventilation, and dust collection.
  • Constructed with a clear, FDA-acceptable PVC wall and internal wire helix reinforcement
  • Operating temperature range of -20°F to 180°F
  • Maximum working pressure of 3 psi at 70°F
  • Vacuum rating of 1.25 in. Hg
  • Highly flexible, sturdy, and suitable for low vacuum applications
  • Moisture and chemical resistant for enhanced durability
Catalog No. NC0209737 Supplier Grainger Supplier No. 3AXZ1
This industrial ducting hose is a flexible, transparent solution designed for the efficient movement of large volumes of air. It features a clear, durable polyvinylchloride (PVC) wall reinforced with a spring wire helix, making it a highly compressible and versatile choice for demanding industrial environments. The hose is well-suited for applications involving chemical fume removal, solvent ventilation, and dust collection.
  • Constructed with a clear, FDA-acceptable PVC wall and internal wire helix reinforcement
  • Operating temperature range of -20°F to 180°F
  • Maximum working pressure of 3 psi at 70°F
  • Vacuum rating of 1.25 in. Hg
  • Highly flexible, sturdy, and suitable for low vacuum applications
  • Moisture and chemical resistant for enhanced durability