Learn More
World Precision Instrument Flexible Implantable Probe, 0.025 inch sensor diameter, 0.005 sec
SDP
Catalog No. 502538738
Encompass
Flexible Teflon microprobes are used for implantation in tissue, in spectrophotometer cuvettes, rectally in neonatal mice, in water baths, PCR thermal cyclers, etc. Also, measure animal rectal temperatures during surgical procedures and pyrogen testing or skin temperature measurement during exercise physiology studies.
Catalog No. 50-253-8738 Supplier World Precision Instrument Supplier No. IT1E
Flexible Teflon microprobes are used for implantation in tissue, in spectrophotometer cuvettes, rectally in neonatal mice, in water baths, PCR thermal cyclers, etc. Also, measure animal rectal temperatures during surgical procedures and pyrogen testing or skin temperature measurement during exercise physiology studies.