Learn More
World Precision Instrument Flexible Implantable Probe, 0.025 inch sensor diameter
SDP
Catalog No. 502538739
Encompass
Flexible Teflon microprobes are used for implantation in tissue, in spectrophotometer cuvettes, rectally in neonatal mice, in water baths, PCR thermal cyclers, etc. Also, measure animal rectal temperatures during surgical procedures and pyrogen testing or skin temperature measurement during exercise physiology studies.
Catalog No. 50-253-8739 Supplier World Precision Instrument Supplier No. IT18EXLONG
Flexible Teflon microprobes are used for implantation in tissue, in spectrophotometer cuvettes, rectally in neonatal mice, in water baths, PCR thermal cyclers, etc. Also, measure animal rectal temperatures during surgical procedures and pyrogen testing or skin temperature measurement during exercise physiology studies.