Learn More
Braintree Scientific Flexible Implantable Probe, .025 dia, constant 0.1 sec
SDP
Catalog No. 501954978
Encompass
Flexible Implantable Probes - IT Series flexible microprobes can be used for tissue implantation, measurement of small volume liquid samples, rectal temperatures in baby mice, etc. Totally sheathed in chemical resistant Teflon. Quite rugged. Max temp. 150C. Isolated. Type # designates needle gauge. (Or one gauge smaller in thin wall). **Sold for research purposes only and not for human use**
Catalog No. 50-195-4978 Supplier Braintree Scientific Supplier No. IT18
Flexible Implantable Probes - IT Series flexible microprobes can be used for tissue implantation, measurement of small volume liquid samples, rectal temperatures in baby mice, etc. Totally sheathed in chemical resistant Teflon. Quite rugged. Max temp. 150C. Isolated. Type # designates needle gauge. (Or one gauge smaller in thin wall). **Sold for research purposes only and not for human use**