Learn More
World Precision Instrument Flexible Light Guide, 18"
SDP
Catalog No. 502539282
Encompass
  • Single Pipe Light Guide with focusing lens
  • Fiber diameter: 6mm
  • 18" (460mm) long, 5/8" standard adapter for attaching to a fiber optic illuminator like the Z-Lite series
Catalog No. 50-253-9282 Supplier World Precision Instrument Supplier No. 504930
  • Single Pipe Light Guide with focusing lens
  • Fiber diameter: 6mm
  • 18" (460mm) long, 5/8" standard adapter for attaching to a fiber optic illuminator like the Z-Lite series