Learn More
World Precision Instrument Flexible Light Guide, 24"
SDP
Catalog No. 502539281
Encompass
  • Single Pipe Light Guide with focusing lens
  • Fiber diameter: 6mm
  • 24" (610mm)  long, 5/8" standard adapter for attaching to a fiber optic illuminator like the Z-Lite series
Catalog No. 50-253-9281 Supplier World Precision Instrument Supplier No. 504931
  • Single Pipe Light Guide with focusing lens
  • Fiber diameter: 6mm
  • 24" (610mm)  long, 5/8" standard adapter for attaching to a fiber optic illuminator like the Z-Lite series