Learn More
Agilent Technologies FLEXIBLE METAL FERRULE, ULTIME
Catalog No. NC2118183
Encompass
Flexible Metal ferrule, UltiMetal Plus, 0.8 mm id, for 0.53 mm id fused silica tubing, 10/pk
This item is not returnable. View return policy
Catalog No. NC2118183 Supplier Agilent Technologies Supplier No. G318827503
Flexible Metal ferrule, UltiMetal Plus, 0.8 mm id, for 0.53 mm id fused silica tubing, 10/pk