Learn More
Electron Microscopy Sciences Cold Embedding Flexible Mold 120 x 60 x 45 mm
SDP
Catalog No. 501929687
Encompass_Preferred
This rectangular flexible mold is made of very flexible transparent silicone rubber which makes it very easy to remove the mounts. The inner sides are polished, so the sides of the mounts get clear. The sides are also straight which makes it easier to fasten the mounts in a specimen holder.
Size: 120 x 60 x 45 mm
Catalog No. 50-192-9687 Supplier Electron Microscopy Sciences Supplier No. 5101265
This rectangular flexible mold is made of very flexible transparent silicone rubber which makes it very easy to remove the mounts. The inner sides are polished, so the sides of the mounts get clear. The sides are also straight which makes it easier to fasten the mounts in a specimen holder.
Size: 120 x 60 x 45 mm