Learn More
Cole-Parmer Digi-Sense Gen-Purp Fle x Thermistor, Vinyl Sheath and Tip, Phono Plug, 10ft Lead
Catalog No. NC1619809 Shop All Cole-Parmer Products
Encompass_Preferred
Thermistor probes offer excellent accuracy over the biological temperature range. Probes are electronically isolated and come with a 10ft lead with a phono plug.
Catalog No. NC1619809 Supplier Cole-Parmer Supplier No. 0849102
Thermistor probes offer excellent accuracy over the biological temperature range. Probes are electronically isolated and come with a 10ft lead with a phono plug.